Drucken

Anti-SMAD5 Rabbit Monoclonal Antibody [clone: MMC-1-104-3]

Lieferant: Abcam
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AB92698-100
ABCAAB92698-100EA 945 EUR
ABCAAB92698-100
Anti-SMAD5 Rabbit Monoclonal Antibody [clone: MMC-1-104-3]
Antikörper
Type: Primary
Antigen: SMAD5 (SMAD family member 5)
Clonality: monoclonal
Clone: MMC-1-104-3
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR