Drucken

Anti-KDM4B Rabbit Polyclonal Antibody

Lieferant: Abgent
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AP11950A
ABGEAP11950AEA 354 EUR
ABGEAP11950A
Anti-KDM4B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KDM4B (lysine (K)-specific demethylase 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR