Drucken

Anti-MLNR Rabbit Polyclonal Antibody

Lieferant: Abgent
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AP12669B
ABGEAP12669BEA 354 EUR
ABGEAP12669B
Anti-MLNR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MLNR (motilin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR