Drucken

Anti-TMPO Rabbit Polyclonal Antibody

Lieferant: Abgent
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

AP14741A
ABGEAP14741AEA 354 EUR
ABGEAP14741A
Anti-TMPO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMPO (thymopoietin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR