Drucken
Anti-HHEX Mouse Monoclonal Antibody (Biotin)
Lieferant: Abnova
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
H00003087-AP41
ABNOH00003087-AP41EA 842 EUR
ABNOH00003087-AP41
Anti-HHEX Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human HHEX.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-HHEX, IgG2b kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-HHEX, IgG2a kappa (50 ?g);
Type:
Antigen: HHEX
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human