Drucken

Anti-TRAF3IP2 Mouse Polyclonal Antibody

Lieferant: Abnova
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

H00010758-B01P
ABNOH00010758-B01PEA 397 EUR
ABNOH00010758-B01P
Anti-TRAF3IP2 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human TRAF3IP2 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TRAF3IP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR