Drucken

Alkaline phosphatase assay kit

Lieferant: ABNOVA
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

KA1642
ABNOKA1642EA 648 EUR
ABNOKA1642
Alkaline phosphatase assay kit
Tests Enzymtests

Alkaline phosphatase assay kit allows the colorimetric kinetic determination of serum alkaline phosphatase activity.


  • High sensitivity and wide linear range: Use 5 µl serum or plasma sample; detection limit is 2 U/L, linear up to 800 U/L
  • Homogeneous and simple procedure: Simple 'mix-and-measure' procedure allows reliable quantitation of ALP activity within 5 minutes
  • Robust and amenable to HTS: All reagents are compatible with high-throughput liquid handling instruments


Applications:
Direct Assays: ALP activity in serum, plasma and other sources
Characterisation and quality control for ALP production
Drug Discovery: High throughput screening for ALP inhibitors and evaluation of ALP inhibitors


Lieferumfang: Kit contains assay buffer 50 ml (pH 10,5), Mg Acetate 1,5 ml (0,2 M), pNPP liquid 600 µl (1 M) and Tartrazine calibrator10 ml.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR