Drucken
BspQ I Restriction Endonuclease
Lieferant: ABNOVA
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
P9390.250UL P9389.50UL
ABNOP9390.250ULEA 498 EUR
ABNOP9390.250UL ABNOP9389.50UL
BspQ I Restriction Endonuclease
Nukleinsäure-Reagenzien Restriktionsenzym
The restriction endonuclease is a restriction endonuclease BspQ I that can recognize specific sites and is produced under GMP standards and expressed recombinantly in Escherichia coli. Restriction endonucleases are widely used in various fields such as gene positioning and cloning. This enzyme cleaves DNA rapidly for efficient gene linearization.