Drucken

Anti-ADORA2 Rat Polyclonal Antibody

Lieferant: Abnova
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

PAB24997
ABNOPAB24997EA 523 EUR
ABNOPAB24997
Anti-ADORA2 Rat Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ADORA2A.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: ADORA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR