Drucken
AccuScript high-fidelity reverse transcriptase
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
600180
AGLS600180EA 484 EUR
AGLS600180
AccuScript high-fidelity reverse transcriptase
Nukleinsäure-Reagenzien RT-PCR und cDNA Reagenzien
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
  • MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
  • High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
  • Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.