Drucken
Qualitative DNA Kits
Lieferant: AGILENT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
DNF-905-K0500 DNF-905-K1000 DNF-910-K0500 DNF-910-K1000 DNF-915-K0500 DNF-915-K1000 DNF-920-K0500 DNF-930-K0500 DNF-930-K1000 DNF-935-K0500 DNF-935-K1000
AGLSDNF-905-K0500EA 470 EUR
AGLSDNF-905-K0500 AGLSDNF-905-K1000 AGLSDNF-910-K0500 AGLSDNF-910-K1000 AGLSDNF-915-K0500 AGLSDNF-915-K1000 AGLSDNF-920-K0500 AGLSDNF-930-K0500 AGLSDNF-930-K1000 AGLSDNF-935-K0500 AGLSDNF-935-K1000
Qualitative DNA Kits
Elektrophorese-Systeme Kapillarelektrophorese-Systeme
DNA fragment analysis for PCR, genotyping, and microsatellite applications.
  • Broad concentration range - all kits have a concentration range of 0.5 to 50 ng/µl
  • Reliable sizing - six different kits to choose from with a sizing range varying from 35 to 20,000 bp
  • Low sample volume - only 2 µl of sample is required, minimising precious sample loss to QC steps
  • Quick separations - electrophoresis times as short as 15 minutes
  • High resolution - separate fragments under 300 bp differing by only 3 bp
The qualitative kits provide automated and accurate sizing with relative quantification of DNA fragments. Common uses include the analysis of PCR amplicons, simple sequence repeats (SSRs)/microsatellites, and genotyping. There are six different kits to choose from based on the size of the DNA or PCR fragments, ranging from 35 to 20,000 bp. Specially formulated gels, markers, and ladders are designed for each kit enabling reliable sizing over a broad range with varying applications. High resolution kits enable the separation of fragments under 300 bp with a 3 bp difference.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.