Drucken

Phleomycin

Lieferant: Apollo Scientific
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

BI3852-100MG BI3852-50MG BI3852-25MG
APOSBI3852-100MGEA 306 EUR
APOSBI3852-100MG APOSBI3852-50MG APOSBI3852-25MG
Phleomycin
Phleomycin

Phleomycin is a glycopeptide antibiotic of the bleomycin family that exhibits in vivo activity against bacteria, eukaryotic microorganisms, plant and animal cells.

Storage Temperature: Gefrierschrank
MDL Number: MFCD00131846
CAS-Nummer: 11006-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR