Drucken

Vancomycin Hydrochlorid

Lieferant: Apollo Scientific
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

BIV0155-1G BIV0155-25G BIV0155-5G
APOSBIV0155-1GEA 29 EUR
APOSBIV0155-1G APOSBIV0155-25G APOSBIV0155-5G
Vancomycin Hydrochlorid
Vancomycin Hydrochlorid

Vancomycin is a glycopeptide antibiotic. It exerts its action by inhibiting the formation of the peptidoglycan polymers of the bacterial cell wall. Often used incombination with cefotaxim or carbenicillin.


  • Potency of 1099 mcg/mg
  • Cell culture tested.

Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
MW: 1485.71 g/mol
Storage Temperature: Kühlschrank
MDL Number: MFCD03613611
CAS-Nummer: 1404-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR