Drucken
Anti-GNAL Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ARP54634_P050
AVIVARP54634_P050EA 401 EUR
AVIVARP54634_P050
Anti-GNAL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNAL (guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now