300l Pre-Sterilized Hamilton CO-RE Typ, Liquid Level Sensing Filter Tip. 96 Tips per Rack, 5 Racks/pack, 12 packs/case 1 * 5.760 ST
Lieferant: AXYGEN
300l Pre-Sterilized Hamilton CO-RE Typ, Liquid Level Sensing Filter Tip. 96 Tips per Rack, 5 Racks/pack, 12 packs/case 1 * 5.760 ST