Drucken
Orbital shaker, benchtop, MaxQ™ HP
Lieferant: BARNSTEAD THERMOLYNE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SHKE416HP-1CE
BARNSHKE416HP-1CEEA 5050 EUR
BARNSHKE416HP-1CE
Orbital shaker, benchtop, MaxQ™ HP
Schüttler Kreisschüttler
MaxQ™ HP benchtop orbital shaker suitable for environmental rooms or on lab benches. It features balancing technology and rugged mechanism make smooth high-capacity 24/7 shaking a reality, an industry-leading platform size, wide-range speed, low heat output motor and spill-resistant design. It features set point security which restricts unauthorised or inadvertent changes to key functions. Equipped with alarm to know the shaker performance.
  • Triple counter-balanced mechanism prevents vibration, allowing the unit to run smoothly with heavy, or out-of-balance loads
  • Microprocessor controller displays run and setpoints simultaneously on a 20-character backlit display
  • Easy programming with icon-based keys and alphanumeric messages
  • Oversized, adjustable suction cup feet minimise vibration
  • Can be used at 4 to 40 °C (maximum 80% relative humidity, non-condensing)
It features low heat 24 V DC motor with moisture resistant electronics provides continuous maintenance-free operation on the benchtop or in a chamber.