Drucken

DSS (Disuccinimidylsuberat)

Lieferant: G Bioscienes
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BC04 BC04-Q
BC04EA 574 EUR
BC04 GENOBC04-Q
DSS (Disuccinimidylsuberat)
DSS (Disuccinimidylsuberat)

Disuccinimidyl suberate Homobifunctional Cross Linker.


  • Ideal for receptor ligand cross-linking and coupling antibody to Protein A or G for immunoprecipitations.
  • OneQuant™ DSS is supplied as 8 vials of 2mg DSS, suitable for preparing for ~100µl 50mM stock solution from each vial.

Formel: C₁₆H₂₀N₂O₈
MW: 368.34 g/mol
MDL Number: MFCD00049059
CAS-Nummer: 68528-80-3

Order Now

Spezifikation Testergebnisse

Spacer Arm (Å) 11.4
Reactive Toward Primary Amines
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR