Drucken
Universal-Füllkanülen
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
305211 305181 303129 303129UK
BDAM305211EA 101 EUR
BDAM305211 BDAM305181 BDAM303129 BDAM303129UK
Universal-Füllkanülen
Injektionsnadeln
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
  • The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
  • The blunt filter is designed for safe ampoule medication preparation
  • BD blunt fill needles have female luer fittings, which mate to male luer fittings
  • BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
  • Natural rubber latex is not part of the material formulation
  • Sterile, singe use
Achtung! Bitte beachten: Note: The BD blunt fill and filter needles are not for skin injection.