Drucken
3-Piece Syringes
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
300188 300186 305502 308062 309649UK 309658UK 309628UK 305502UK 301231UK 303179 303174 303173
613-3913EA 0 EUR
613-3913 BDAM300186 BDAM305502 613-1253 BDAM309649UK BDAM309658UK 613-2997 613-2996 BDAM301231UK BDAM303179 BDAM303174 BDAM303173
3-Piece Syringes
Spritzen Spritzen, medizinisch
BD Plastipak™ syringe is a precision drug delivery device that is an essential part of your daily patient care toolkit, which you can use across a range of critical applications and departments. It is intended for general purpose injection and aspiration of fluids from vials, ampoules and parts of the body below the surface of the skin.
  • 3-piece design with latex-free stopper
  • Clear barrel for visualization of syringe content
  • Bold scale printing to aid in setting for a more accurate dosage of medication
  • Special silicone lubrication ensures that plunger moves smoothly and evenly
  • Retaining ring to prevent accidental plunger rod withdrawal
  • Leak-tight
Latex-free stopper prevents leakage of medication around the plunger and acts as an indicator for measuring the syringe contents.
BD Plastipak™, 50 ml dosage:
- Perfusion syringes, 50 ml syringes, are designed for short term use in syringe pumps (active IIa devices) for the administration of pharmaceuticals
- The 50 mL Catheter Tip Syringes have a long tapered tip designed to aid in irrigation or for connection to non-ISO compatible Luer connections such as nasogastric tubes
BD Plastipak™, 1 ml low dosage:
- Low dose syringes make it easier to accurately measure a small dose than with large capacity syringes
- Available without or with needles (25G, 26G, 27G or 30G)
- 1 ml syringes with 0,01 ml, I.U 40 or I.U 100 graduations
- The BD Plastipak™ Test and Sub-Q syringes are mainly designed for administering allergens in the desensitization treatment of allergies