Drucken
BD Plastipak™ Syringes
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
300605 300613 300629 300865 300866 300912 305501 301029 301073 300869 301035 300328 301027 309628 301229 301231 300867 309653 309649 309658 302830 302237 309648 302238 303172 303175 301031 305959 300223
613-3920EA 112 EUR
613-3920 613-3921 613-3922 613-3925 613-3926 613-3931 613-2989 BDAM301029 BDAM301073 615-0033 BDAM301035 BDAM300328 BDAM301027 613-4971 BDAM301229 612-3677 613-1124 BDAM309653 613-1997 613-1998 BDAM302830 BDAM302237 BDAM309648 BDAM302238 613-5399 613-5413 BDAM301031 BDAM305959 BDAM300223
BD Plastipak™ Syringes
Spritzen Spritzen, medizinisch
BD Plastipak™ Three-Piece Syringe is a precision drug delivery device that is an essential part of your daily patient care toolkit, which you can use across a range of critical applications and departments. It is intended for general purpose injection and aspiration of fluids from vials, ampoules and parts of the body below the surface of the skin.
  • Clear barrel for visualisation of syringe content
  • Bold scale printing to aid in setting a more accurate medication dosage
  • Special silicone lubrication ensures the plunger moves smoothly and evenly
  • Retaining ring to prevent accidental plunger rod withdrawal
  • Leak-tight
  • Natural rubber latex is not part of the material formulation
  • Sterile (also available as non sterile), single-use
Latex-free stopper prevents leakage of medication around the plunger and acts as an indicator for measuring the syringe contents.