Drucken
Insulinspritzen, BD Micro-Fine™
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
320926 320927 324892 324891 324826 324825 324827 324824
BDAM320926EA 155 EUR
BDAM320926 BDAM320927 BDAM324892 BDAM324891 BDAM324826 BDAM324825 BDAM324827 BDAM324824
Insulinspritzen, BD Micro-Fine™
Spritzen Spritzen, medizinisch
Micro-Fine™ is a single-use syringe for diabetes. It offers a more comfortable and convenient way to inject insulin with three different volumes.
  • Made from surgical stainless steel
  • Precision and comfort
  • Optimum shape and sharpness of the needle tip and bevel
  • Electro polished needle shaft
The white and orange protective cap conforms with sterility and a minimal dead space reduces the risk of air bubbles and prevents waste of insulin.