Drucken
Plasma separator tubes, PST™ II, BD Vacutainer®
Lieferant: BECTON DICKINSON MEDICAL
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
367376 367374 367378
BDAM367376EA 106 EUR
BDAM367376 BDAM367374 BDAM367378
Plasma separator tubes, PST™ II, BD Vacutainer®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
Plasma tubes with separating gel for clinical chemistry are available with spray-dried lithium heparin additives.
  • It act as stable barrier between plasma and cells providing analyte stability and better sample quality
  • Correct mixing of the tubes immediately after the blood sample collection is important to avoid micro clotting
  • Plasma separation tubes should be gently inverted 180° and back 8 to 10 times
During the centrifugation of these tubes, an inert gel separates the plasma and the cells preventing the contamination of the plasma from the separated cellular components.
Bestellinformation: Study documentation in relation to BD Vacutainer® PST™ II tubes are supplied on request.