Drucken
Environmentally friendly thermometer, individually calibrated, Enviro-Safe®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
B602020100
BELAB602020100EA 439 EUR
BELAB602020100
Environmentally friendly thermometer, individually calibrated, Enviro-Safe®, SP Bel-Art
Thermometer Analoge Thermometer
Each easy to read thermometer includes an individualised thermometer calibration report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • EnviroKleen™ certified
  • Non toxic, Enviro-Safe® liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors
Each instrument is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology.
Zertifizierungen: Traceable to NIST, individually serialised.