Drucken
Markers, bel-pen, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
F133740000
BELAF133740000EA 61 EUR
BELAF133740000
Markers, bel-pen, SP Bel-Art
Schreibstifte Permanentmarker
These felt tip marking pens are perfect for labeling all types of glassware and plastics. The permanent and quick drying ink remains waterproof and smudge proof.
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 133 mm long