Drucken
Desiccator cabinets, mini, Secador®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
F420751000
BELAF420751000EA 414 EUR
BELAF420751000
Desiccator cabinets, mini, Secador®, SP Bel-Art
Exsikkatoren Trockenkammern
This dry storage cabinet is perfect for small, humidity sensitive products or materials that are always needed close to a work area.
  • Large side opening door maximises access to interior space
  • Easy-to-view dial hygrometer located on the door front
  • Can be used with desiccant cartridges 467-0000 and 467-0001
  • Kit available to mount to unit on the underside of a shelf
Cabinet comprises of clear polystyrene and door is made from clear co-polyester which makes viewing contents easy. A positive latch on the door provides an excellent seal and two locking loops allow a padlock or tamper evident seal to be used to maintain security.
Achtung! Bitte beachten: Mini desiccator model does not block UV light.