Drucken
Digital refractometer, RFM700-M
Lieferant: BELLINGHAM STANLEY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
19-00-61-181 19-00-61-182 19-00-61-191 19-10-61-181 19-10-61-182 19-10-61-191 19-00 19-10
BELL19-00-61-181EA 5990 EUR
BELL19-00-61-181 BELL19-00-61-182 BELL19-00-61-191 BELL19-10-61-181 BELL19-10-61-182 BELL19-10-61-191 BELL19-00 BELL19-10
Digital refractometer, RFM700-M
Refraktometer Tischrefraktometer
Robust fully automatic instruments for the food, sugar and beverage industries where temperature control is not required, such as measuring sugar (°Brix) concentrations in the food processing industry.
  • Automatic sense 'hands free' measurements
  • Simple audit trail with date, time and batch number
  • Alphanumeric keypad for easy data entry
  • Flat sapphire prism surface for easy-cleaning
  • Selectable read type: Continual, single, intelligent auto-sense
Designed with ultimate simplicity in mind, the RFM700-M refractometers featuring automatic temperature compensation offer digital automatic technology. Latest models feature more scale and temperature correction options, enabling the instrument to be used in the traditional food, sugar and beverage applications but also in a number of other ”non-sucrose” based industrial applications.
Particularly suited to wine applications, as industry standard scales such as Baume, Oechsle, and Alcohol Probable (A.P.) are all accessible from the on-board library.
Zertifizierungen: ASTM D 5006 - ICUMSA (Internationale Kommission für einheitliche Methoden der Zuckeranalyse).
Lieferumfang: Supplied with relevant cable for power supply (100 - 240 V, 50 - 60 Hz) and multi-language instruction manual. EU, UK and US plugs are supplied with the unit, CH and Danish plugs must be ordered separately.