Drucken
Digital refractometers, RFM300-T
Lieferant: BELLINGHAM STANLEY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
19-30-61-181 19-30-61-182 19-30-61-191 19-40-61-181 19-40-61-182 19-40-61-191 19-30 19-40
635-0730EA 12190 EUR
635-0730 635-0731 635-0732 635-0734 635-0735 635-0736 BELL19-30 BELL19-40
Digital refractometers, RFM300-T
Refraktometer Tischrefraktometer
These high precision refractometers are designed for applications in food and beverage, including fruit juice, emulsions or chemicals.
  • Flat prism surface, touch screen and Peltier control
  • Smart temperature stability check only displays results when sample is stable
  • Up to 8000 readings stored in memory can be downloaded to a PC using free downloadable software
Zertifizierungen: US/EP/BP/JP. FDA 21 CFR part 11. Validation documentation IQ/OQ/PQ is available on request.
Lieferumfang: Supplied with power cable and multi-language CD manual. EU, UK and US plugs are supplied with the unit, CH and Danish plugs must be ordered separately.