Drucken
Torso
Lieferant: Binhold, Paul Lehrmittel
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
1005618 1008541
BINH1005618EA 0 EUR
BINH1005618 BINH1008541
Torso
Modelle Humanbiologie
This easy to transport torso allows the practice of adult CPR techniques. Optionally child CPR can also be practiced by changing the supplied springs inside the torso. Individual student face masks with a one-way non-rebreathing valve and disposable airways provide maximum protection against cross-contamination.
  • With light controller