Drucken

(S)-(-)-β-Citronellol 95%

Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BD218718-500G
BLDPBD218718-500GEA 652 EUR
BLDPBD218718-500G
(S)-(-)-β-Citronellol 95%
(S)-(-)-β-Citronellol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00063214
CAS-Nummer: 7540-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR