Drucken

Anti-EXPB-2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-0243R-A680
BOSSBS-0243R-A680EA 890 EUR
BOSSBS-0243R-A680
Anti-EXPB-2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
beta-expansin (EXBP2) may cause loosening and extension of plant cell walls by disrupting noncovalent bonding between cellulose microfibrils and matrix glucans. No enzymatic activity has been found Subcellular Location at Cell-wall bound. Belongs to the expansin family.

Type: Primary
Antigen: EXPB-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 10-110/265
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR