Drucken
Anti-EXPB-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-0243R-A750
BOSSBS-0243R-A750EA 890 EUR
BOSSBS-0243R-A750
Anti-EXPB-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
beta-expansin (EXBP2) may cause loosening and extension of plant cell walls by disrupting noncovalent bonding between cellulose microfibrils and matrix glucans. No enzymatic activity has been found Subcellular Location at Cell-wall bound. Belongs to the expansin family.
Type: Primary
Antigen: EXPB-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 10-110/265
Host: Rabbit
Isotype: IgG
Reactivity: Plant