Drucken

Anti-GRN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-0823R-FITC
BOSSBS-0823R-FITCEA 909 EUR
BOSSBS-0823R-FITC
Anti-GRN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Granulins have possible cytokine-like activity. They may play a role in inflammation, wound repair, and tissue remodeling. Granulin-4 promotes proliferation of the epithelial cell line A431 in culture while granulin-3 acts as an antagonist to granulin-4, inhibiting the growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Granulin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR