Drucken
Anti-GLI1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-1206R-A680
BOSSBS-1206R-A680EA 899 EUR
BOSSBS-1206R-A680
Anti-GLI1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Acts as a transcriptional activator. May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.
Type: Primary
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-700/1106
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig