Drucken

Anti-GLI1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-1206R-CY5.5
BOSSBS-1206R-CY5.5EA 909 EUR
BOSSBS-1206R-CY5.5
Anti-GLI1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Acts as a transcriptional activator. May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR