Drucken
Anti-YWHAQ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-12419R-HRP
BOSSBS-12419R-HRPEA 900 EUR
BOSSBS-12419R-HRP
Anti-YWHAQ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathways. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner. Negatively regulates the kinase activity of PDPK1.This antibody is reactive with 14-3-3 Alpha/Beta/Gamma/Delta/Epsilon.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: YWHAQ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat