Drucken

Anti-APOA1BP Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-12491R-A647
BOSSBS-12491R-A647EA 893 EUR
BOSSBS-12491R-A647
Anti-APOA1BP Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Apolipoproteins are protein components of plasma lipoproteins. Apolipoprotein A-I (ApoA1) promotes cholesterol efflux from tissues to the liver for excretion. ApoA1 is the major protein component of high density lipoprotein (HDL) in the plasma. It can function as a cofactor for lecithin cholesterolacyltransferase, which is responsible for the formation of most plasma cholesteryl esters. AI-BP (Apolipoprotein A-I-binding protein), also known as YjeF N-terminal domain-containing protein 1, is a 288 amino acid secreted protein that binds ApoA1, ApoA2 and HDL. Individuals with impaired renal function show an increased rate of AI-BP excretion, indicating that it is normally reabsorbed within the kidney tubules. AI-BP belongs to the YjeF N-terminal domain protein family, which includes proteins that are frequently involved in oogenesis and spermatogenesis. There are two isoforms of AI-BP that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOA1BP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR