Drucken

Anti-CRYBA1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-12859R-A488
BOSSBS-12859R-A488EA 893 EUR
BOSSBS-12859R-A488
Anti-CRYBA1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Crystallins are the major proteins of the vertebrate eye lens, where they maintain the transparency and refractive index of the lens. Crystallins are divided into a, b, and g families, and the b- and g-crystallins also comprise a superfamily. Crystallins usually contain seven distinctive protein regions, including four homologous motifs, a connecting peptide, and N- and C-terminal extensions. b-crystallins constitute the major lens structural proteins. They associate into dimers, tetramers, and higher order aggregates. The b-crystallin subfamily is composed of several gene products, including bA1-, bA2-, bA3-, bA4-, bB1-, bB2- and bB3-crystallin. The bA1- and bA3-crystallin proteins are encoded by a single mRNA. They differ by only 17 amino acids, and bA1-crystallin is generated by use of an alternate translation initiation site. The genes for bA4-, bB1-, bB2- and bB3-crystallin are clustered on human chromosome 22q11, while the genes for bA3/A1- and bA2-crystallin map to human chromosomes 17q11 and 2q34, respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRYBA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR