Drucken
Anti-PCDHB6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-13726R-A680
BOSSBS-13726R-A680EA 890 EUR
BOSSBS-13726R-A680
Anti-PCDHB6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHB6 (Protocadherin beta-6) is a 794 amino acid single pass transmembrane protein that is one of 16 proteins in the protocadherin beta cluster. Unlike the alpha and gamma gene clusters whose genes are spliced to downstream constant region exons during transcription, members of the beta cluster (such as PCDHB6) do not use constant-region exons to produce mRNAs. As a result, each protocadherin beta gene encodes the transmembrane, extracellular and short cytoplasmic domains of the protein. PCDHB6 is likely a calcium-dependent cell adhesion protein that is involved in the maintenance of neural connections in the brain. Unlike most protocadherin-beta proteins, PCDHB6 has not one but two PXXP motifs within its cytoplasmic domain, suggesting a role in signal transduction cascade events.
Type: Primary
Antigen: PCDHB6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/794
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now