Drucken

Anti-PCDHB14 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-13727R-A647
BOSSBS-13727R-A647EA 893 EUR
BOSSBS-13727R-A647
Anti-PCDHB14 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHB14 (Protocadherin beta 14) is a 798 amino acid protein that is one of 16 proteins in the protocadherin beta cluster. Unlike the alpha and gamma gene clusters whos genes are spliced to downstream constant region exons during transcription, members of the beta cluster (such as PCDHB14) do not use constant-region exons to produce mRNAs. As a result, each protocadherin beta gene encodes the transmembrane, extracellular and short cytoplasmic domains of the protein. Localized to the cell membrane, PCDHB14 is a single-pass type I membrane protein that contains six cadherin domains.

Type: Primary
Antigen: PCDHB14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 551-650/798
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR