Drucken

Anti-beta Adaptin/AP2 beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-1546R-A680
BOSSBS-1546R-A680EA 890 EUR
BOSSBS-1546R-A680
Anti-beta Adaptin/AP2 beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. AP2 beta appears to be required for normal face and limb development and for proper terminal differentiation and function of renal tubular epithelia.

Type: Primary
Antigen: beta Adaptin/AP2 beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-450/937
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR