Drucken
Anti-LCAT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-1972R-A488
BOSSBS-1972R-A488EA 893 EUR
BOSSBS-1972R-A488
Anti-LCAT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Central enzyme in the extracellular metabolism of plasma lipoproteins. Synthesized mainly in the liver and secreted into plasma where it converts cholesterol and phosphatidylcholines (lecithins) to cholesteryl esters and lysophosphatidylcholines on the surface of high and low density lipoproteins (HDLs and LDLs). The cholesterol ester is then transported back to the liver. Has a preference for plasma 16:0-18:2 or 18:O-18:2 phosphatidylcholines. Also produced in the brain by primary astrocytes, and esterifies free cholesterol on nascent APOE-containing lipoproteins secreted from glia and influences cerebral spinal fluid (CSF) APOE- and APOA1 levels. Together with APOE and the cholesterol transporter ABCA1, plays a key role in the maturation of glial-derived, nascent lipoproteins. Required for remodeling high-density lipoprotein particles into their spherical forms.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LCAT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat