Drucken

Anti-Methionine adenosyltransferase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-4054R-A750
BOSSBS-4054R-A750EA 874 EUR
BOSSBS-4054R-A750
Anti-Methionine adenosyltransferase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
S adenosylmethionine synthetase catalyses the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.

Type: Primary
Antigen: Methionine adenosyltransferase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/395
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR