Drucken

Anti-TM7SF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-4221R-A750
BOSSBS-4221R-A750EA 890 EUR
BOSSBS-4221R-A750
Anti-TM7SF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Involved in the conversion of lanosterol to cholesterol.Expressed in adult heart, brain, pancreas, lung, liver, skeletal muscle, kidney, ovary, prostate, and testis, but not detected in placenta, spleen, thymus, small intestine, colon (mucosal lining), or peripheral blood leukocytes.

Type: Primary
Antigen: TM7SF2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/418
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR