Drucken
Anti-CKS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-5745R-A750
BOSSBS-5745R-A750EA 890 EUR
BOSSBS-5745R-A750
Anti-CKS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
CKS1 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein.
Type: Primary
Antigen: CKS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-79/79
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now