Drucken
Anti-KLHL7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BS-5935R-A488
BOSSBS-5935R-A488EA 893 EUR
BOSSBS-5935R-A488
Anti-KLHL7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin ligase complex. The BCR(KLHL7) complex acts by mediating ubiquitination and subsequent degradation of substrate proteins. Probably mediates 'Lys-48'-linked ubiquitination.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLHL7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat