Drucken

Anti-REG3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-6291R-A488
BOSSBS-6291R-A488EA 893 EUR
BOSSBS-6291R-A488
Anti-REG3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Phosphatase enzymes catalyse hydrolysis of phosphoric acid esters of various alcohols, e.g. a hexose phosphate, to yield the alcohol and free inorganic phosphate. This may be a way of recycling phosphate in the cell, and the level of phosphate may be elevated under conditions of phosphate starvation (in algae for example). Acid phosphatases have pH optima below 7, whereas alkaline phosphatases are most active above pH 7. Commercial quantities of acid phosphatase are obtained from plant sources e.g. potato and wheat germ, whereas similar quantities of alkaline phosphatase are obtained from animal intestine and bacteria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: REG3A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR