Drucken

Anti-Acid Phosphatase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-6291R-A750
BOSSBS-6291R-A750EA 893 EUR
BOSSBS-6291R-A750
Anti-Acid Phosphatase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Phosphatase enzymes catalyse hydrolysis of phosphoric acid esters of various alcohols, e.g. a hexose phosphate, to yield the alcohol and free inorganic phosphate. This may be a way of recycling phosphate in the cell, and the level of phosphate may be elevated under conditions of phosphate starvation (in algae for example). Acid phosphatases have pH optima below 7, whereas alkaline phosphatases are most active above pH 7. Commercial quantities of acid phosphatase are obtained from plant sources e.g. potato and wheat germ, whereas similar quantities of alkaline phosphatase are obtained from animal intestine and bacteria.

Type: Primary
Antigen: Acid Phosphatase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 71-158/158
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR