Drucken

Anti-UHRF1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-6427R-A647
BOSSBS-6427R-A647EA 915 EUR
BOSSBS-6427R-A647
Anti-UHRF1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Putative E3 ubiquitin-protein ligase. May participate in methylation-dependent transcriptional regulation. Binds to inverted 5'-CCAAT-3' box 2 in the TOP2A promoter, and activates TOP2A expression. Important for G1/S transition. May be involved in DNA repair and chromosomal stability.Tissue specificity:Expressed in thymus, bone marrow, testis, lung and heart. Overexpressed in breast cancer. UHRF2 is an E3 ubiquitin-protein ligase that is an intermolecular hub protein in the cell cycle network. Through cooperative DNA and histone binding, may contribute to a tighter epigenetic control of gene expression in differentiated cells. Ubiquitinates cyclins, CCND1 and CCNE1, in an apparently phosphorylation-independent manner and induces G1 arrest. Also ubiquitinates PCNP leading to its degradation by the proteasome. E3 SUMO-, but not ubiquitin-, protein ligase for ZNF131.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UHRF1/2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR