Drucken

Anti-YWHAZ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-6790R-FITC
BOSSBS-6790R-FITCEA 900 EUR
BOSSBS-6790R-FITC
Anti-YWHAZ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathways. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: YWHAZ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR