Drucken

Anti-ADGRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

BS-7513R-A555
BOSSBS-7513R-A555EA 893 EUR
BOSSBS-7513R-A555
Anti-ADGRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
BAI 1 protein is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain. Reduced or no expression is observed in some glioblastoma cell lines and cancer tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR